2-Phenoxyethyl acetate ≥99.0%

Supplier: TCI America

A0935-25G A0935-500G
TCA0935-25GEA 50.56 CAD
TCA0935-25G TCA0935-500G
2-Phenoxyethyl acetate ≥99.0%
2-Phenoxyethyl acetate

CAS Number: 6192-44-5
MDL Number: MFCD00059336
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Flash Point (°C): 141
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: Acetic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Acetate

Formula: C₁₀H₁₂O₃
Boiling Pt: 231 °C
Density: 1.11 (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00059336
CAS Number: 6192-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=454a32bd051ef20c08dd6dfbbaa38ca16e2d8b9d&vid=454a32bd051ef20c08dd6dfbbaa38ca16e2d8b9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR