4,5-Dimethyl-o-phenylenediamine ≥98.0% (by HPLC)

Supplier: TCI America

D1083-25G D1083-5G
TCD1083-025GEA 357.08 CAD
TCD1083-025G TCD1083-5G
4,5-Dimethyl-o-phenylenediamine ≥98.0% (by HPLC)
4,5-Dimethyl-o-phenylenediamine

CAS Number: 3171-45-7
MDL Number: MFCD00007729
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 127


Other Product Names / Synonyms: 4,5-Diamino-o-xylene

Formula: C₈H₁₂N₂
Melting Pt: 127 °C
MDL Number: MFCD00007729
CAS Number: 3171-45-7
EINECS: 221-635-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d72cece97973a138d42d8cd23c2b3d2c0dcd9926&vid=d72cece97973a138d42d8cd23c2b3d2c0dcd9926&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR