Decyl decanoate ≥95.0%

Supplier: TCI America

D1080-25ML D1080-500ML
TCD1080-025MLEA 115.34 CAD
TCD1080-025ML TCD1080-500ML
Decyl decanoate ≥95.0%
Decyl decanoate

CAS Number: 1654-86-0
MDL Number: MFCD00059280
Molecular Formula: C20H40O2
Molecular Weight: 312.54
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 159
Flash Point (°C): 140
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Decanoic Acid Decyl Ester

Formula: C₂₀H₄₀O₂
MW: 312.5304 g/mol
Boiling Pt: 159 °C
Density: 0.86 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00059280
CAS Number: 1654-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c1690c315cfc33240f846c1a7879cf786e88403&vid=9c1690c315cfc33240f846c1a7879cf786e88403&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR