Oleyl acetate 60,0%

Supplier: TCI America

Synonyms: Acetic Acid Oleyl Ester

A0934-500ML A0934-25ML
TCA0934-500MLEA 436.08 CAD
TCA0934-500ML TCA0934-025ML
Oleyl acetate 60,0%
Oleyl acetate

CAS Number: 693-80-1
MDL Number: MFCD00056326
Molecular Formula: C20H38O2
Molecular Weight: 310.52
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 186
Flash Point (°C): 190
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid Oleyl Ester

Formula: C₂₀H₃₈O₂
Boiling Pt: 186 °C
Density: 0.87 (20 °C)
Flash Pt: 190 °C
MDL Number: MFCD00056326
CAS Number: 693-80-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d126593517bb27d75dd2802185d87750ffae6ec&vid=2d126593517bb27d75dd2802185d87750ffae6ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 60.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR