2,4-Diamino-6-diallylamino-1,3,5-triazine ≥98.0%

Supplier: TCI America

Synonyms: N2,N2-Diallylmelamine

D1075-25G D1075-500G
TCD1075-025GEA 132.72 CAD
TCD1075-025G TCD1075-500G
2,4-Diamino-6-diallylamino-1,3,5-triazine ≥98.0%
2,4-Diamino-6-diallylamino-1,3,5-triazine

CAS Number: 91-77-0
MDL Number: MFCD00047347
Molecular Formula: C9H14N6
Molecular Weight: 206.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 145


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: N2,N2-Diallylmelamine

Formula: C₉H₁₄N₆
MW: 206.25 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00047347
CAS Number: 91-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7287ac36fae9a6d725968ff69019605dd770bd2&vid=e7287ac36fae9a6d725968ff69019605dd770bd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR