Dithiothreitol (DTT, Cleland's reagent) 98,0 for synthesis

Supplier: TCI America
Warning

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

D1071-1G D1071-5G D1071-25G
TCD1071-001GEA 76.98 CAD
TCD1071-001G TCD1071-005G TCD1071-25G
Dithiothreitol (DTT, Cleland's reagent) 98,0 for synthesis
Dithiothreitol (DTT, Cleland's reagent)

CAS Number: 3483-12-3
MDL Number: MFCD00004877
Molecular Formula: C4H10O2S2
Molecular Weight: 154.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 43
Flash Point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Bioscience, Sugar Alcohols, Sugars (Others), O-Substituted Sugars, Functionality Creating Reagents, Disulfide Reducing Agents
Other Product Names / Synonyms: Cleland's Reagent, DL-2,3-Dihydroxy-1,4-butanedithiol, DL-1,4-Dimercapto-2,3-butanediol, DL-DTT

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 130 °C
Melting Pt: 43 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd1523edc87c93981d015610c18a5add7a94a23&vid=0bd1523edc87c93981d015610c18a5add7a94a23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR