Dipropyleneglycol monomethyl ether (mixture of isomers) ≥98.0%

Supplier: TCI America

Synonyms: Dipropylene glycol monomethyl ether, 1-((1-Methoxypropan-2-yl)oxy)propan-2-ol

D1053-25G D1053-500G
TCD1053-025GEA 28.44 CAD
TCD1053-025G TCD1053-500G
Dipropyleneglycol monomethyl ether (mixture of isomers) ≥98.0%
Dipropyleneglycol monomethyl ether (mixture of isomers)

CAS Number: 34590-94-8
MDL Number: MFCD00059604
Molecular Formula: C7H16O3
Molecular Weight: 148.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Melting point (°C): -80
Flash Point (°C): 77
Specific Gravity (20/20): 0.95

Formula: C₇H₁₆O₃
MW: 148.2001 g/mol
Boiling Pt: 188 °C
Melting Pt: –80 °C
Density: 0.95 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00059604
CAS Number: 34590-94-8
EINECS: 252-104-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f903db8ad2670c203820d1bff6a2bf10b568540&vid=8f903db8ad2670c203820d1bff6a2bf10b568540&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR