Homogentisic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
D1050-1G
D1050-5G
D1050-200MG
TCD1050-001GEA
455.04
CAD
TCD1050-001G
TCD1050-005G
TCD1050-200MG
Homogentisic acid ≥97.0% (by HPLC, titration analysis)
Homogentisic acid
CAS Number: 451-13-8
MDL Number: MFCD00004324
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 147
Other Product Names / Synonyms: 2,5-Dihydroxyphenylacetic Acid
Formula:
C₈H₈O₄ Melting Pt: 147 °C |
MDL Number:
MFCD00004324 CAS Number: 451-13-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...