3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0%

Supplier: TCI America

Synonyms: 3,9-Divinylspirobi(m-dioxane)

D1027-500G D1027-25G D1027-100G
TCD1027-500GEA 1294.02 CAD
TCD1027-500G TCD1027-025G TCD1027-100G
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0%
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane

CAS Number: 78-19-3
MDL Number: MFCD00006564
Molecular Formula: C11H16O4
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 94
Melting point (°C): 44
Flash Point (°C): 110


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3,9-Divinylspirobi(m-dioxane)

Formula: C₁₁H₁₆O₄
MW: 212.25 g/mol
Boiling Pt: 94 °C
Melting Pt: 44 °C
Flash Pt: 110 °C
MDL Number: MFCD00006564
CAS Number: 78-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cee2249056a9d2fad1513f65332cf517b3208bf3&vid=cee2249056a9d2fad1513f65332cf517b3208bf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR