2,3-Dichlorotetrahydrofuran ≥90.0%

Supplier: TCI America

Synonyms: Tetrahydro-2,3-dichlorofuran

D0985-25G D0985-5G
TCD0985-025GEA 591.71 CAD
TCD0985-025G TCD0985-5G
2,3-Dichlorotetrahydrofuran ≥90.0%
2,3-Dichlorotetrahydrofuran

CAS Number: 3511-19-1
MDL Number: MFCD00059736
Molecular Formula: C4H6Cl2O
Molecular Weight: 140.99
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Specific Gravity (20/20): 1.34


Other Product Names / Synonyms: Tetrahydro-2,3-dichlorofuran

Formula: C₄H₆Cl₂O
MW: 140.99 g/mol
Boiling Pt: 63 °C
Density: 1.34 (20 °C)
MDL Number: MFCD00059736
CAS Number: 3511-19-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bbdaf63223b63664610043488e6cf38b7b625b3&vid=3bbdaf63223b63664610043488e6cf38b7b625b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR