Lauroylcholine chloride hydrate ≥98.0%

Supplier: TCI America

Synonyms: Dodecanoylcholine Chloride

D0973-1G D0973-5G
TCD0973-001GEA 178.4 CAD
TCD0973-001G TCD0973-5G
Lauroylcholine chloride hydrate ≥98.0%
Lauroylcholine chloride hydrate

CAS Number: 25234-60-0
MDL Number: MFCD00050185
Molecular Formula: C17H36ClNO2
Molecular Weight: 321.93
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Dodecanoylcholine Chloride

Formula: C₁₇H₃₆ClNO₂
MDL Number: MFCD00050185
CAS Number: 25234-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a45e918a4b2dccc192334e3236a72fa01ad620ac&vid=a45e918a4b2dccc192334e3236a72fa01ad620ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR