4,4'-Dimethyldiphenylamine ≥98.0%
Supplier: TCI America
D0950-25G
D0950-5G
TCD0950-025GEA
144.66
CAD
TCD0950-025G
TCD0950-005G
4,4'-Dimethyldiphenylamine ≥98.0%
4,4'-Dimethyldiphenylamine
CAS Number: 620-93-9
MDL Number: MFCD00059317
Molecular Formula: C14H15N
Molecular Weight: 197.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 330
Melting point (°C): 79
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Diphenylamines (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Dimethyldiphenylamine
Formula:
C14H15N Boiling Pt: 330 °C Melting Pt: 79 °C |
MDL Number:
MFCD00059317 CAS Number: 620-93-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...