2-Amino-2-oxazoline hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A0923-5G
A0923-25G
TCA0923-005GEA
491.38
CAD
TCA0923-005G
TCA0923-25G
2-Amino-2-oxazoline hydrochloride ≥98.0% (by titrimetric analysis)
2-Amino-2-oxazoline hydrochloride
CAS Number: 375855-07-5
MDL Number: MFCD00060178
Molecular Formula: C3H6N2O
Molecular Weight: 122.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 108
Formula:
C₃H₆N₂O Melting Pt: 108 °C |
MDL Number:
MFCD00060178 CAS Number: 375855-07-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...