Dipropyleneglycol (mixture of isomers) ≥95.0%
Supplier: TCI America
Synonyms:
Dipropylene glycol
D0933-25G
D0933-500G
TCD0933-25GEA
37.92
CAD
TCD0933-25G
TCD0933-500G
Dipropyleneglycol (mixture of isomers) ≥95.0%
Dipropyleneglycol (mixture of isomers)
CAS Number: 25265-71-8
MDL Number: MFCD00004538
Molecular Formula: C6H14O3
Molecular Weight: 134.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 138
Specific Gravity (20/20): 1.03
Formula:
C₆H₁₄O₃ MW: 134.18 g/mol Density: 1.03 (20 °C) Flash Pt: 138 °C |
MDL Number:
MFCD00051023 CAS Number: 25265-71-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...