Diisopropylamine hydrochloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America

D0926-25G D0926-500G
TCD0926-025GEA 90.06 CAD
TCD0926-025G TCD0926-500G
Diisopropylamine hydrochloride ≥99.0% (by titrimetric analysis)
Diisopropylamine hydrochloride

CAS Number: 819-79-4
MDL Number: MFCD00034860
Molecular Formula: C6H15N
Molecular Weight: 137.65
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 217

Formula: C₆H₁₅N
Melting Pt: 217 °C
MDL Number: MFCD00034860
CAS Number: 819-79-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05d88e7deb4ad202bc19303e901164f4e2fa7422&vid=05d88e7deb4ad202bc19303e901164f4e2fa7422&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR