β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate, white, crystals for biochemistry
Supplier: TCI America
|
Warning
|
Synonyms:
beta-NADH disodium salt hydrate, beta-Nicotinamide adenine dinucleotide disodium salt hydrate, β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate
D0920-1G
D0920-100MG
D0920-5G
TCD0920-001GEA
279.77
CAD
TCD0920-001G
TCD0920-100MG
TCD0920-5G
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate, white, crystals for biochemistry
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate
CAS Number: 606-68-8
Molecular Formula: C21H29N7O14P2
Molecular Weight: 709.41
Form: Crystals
Color: White
Product Categories: BioscienceNucleosides, Nucleotides, Nucleic Acids, Reagents for Bioanalytical Research, Vitamins
Formula:
C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O MW: 727.43 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036200 CAS Number: 606-68-8 EINECS: 210-123-3 |
Specification Test Results
Storage Temperature |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...