β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, white ≥95%, crystals for biochemistry

Supplier: TCI America

Synonyms: Cozymase, beta-DPN, Coenzyme I, beta-Nicotinamide Adenine dinucleotide hydrate, oxidized form, beta-Diphosphopyridine Nucleotide, beta-NAD

D0919-1G D0919-5G
TCD0919-001GEA 135 CAD
TCD0919-001G TCD0919-005G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) hydrate, white ≥95%, crystals for biochemistry
β-Nicotinamide adenine dinucleotide (NAD, oxidized form) hydrate

CAS Number: 53-84-9
Molecular Formula: C21H27N7O14P2
Molecular Weight: 663.43
Form: Crystals
Color: White


Product Categories: BioscienceNucleosides, Nucleotides, Nucleic Acids, Reagents for Bioanalytical Research, Vitamins

Formula: C₂₁H₂₇N₇O₁₄P₂·H₂O
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)
Storage Temperature 0-10°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR