Potassium-L-aspartate hydrate 97,0 Titrimetric

Supplier: TCI America

Synonyms: H-Asp-OH(Potasium), L-Asparaginic acid Potassium salt hydrate, L-aspartic acid potasium salt, L-Aspartic acid Potassium salt hydrate

A0922-25G A0922-500G
TCA0922-025GEA 80.5 CAD
TCA0922-025G TCA0922-500G
Potassium-L-aspartate hydrate 97,0 Titrimetric
Potassium-L-aspartate hydrate

CAS Number: 1115-63-5
MDL Number: MFCD00021083
Molecular Formula: C4H7NO4
Molecular Weight: 171.19
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Specific rotation [a]20/D: 20.5 deg (C=8, 6mol/L HCl)


Other Product Names / Synonyms: L-Asparaginic Acid Potassium Salt, L-Aspartic Acid Potassium Salt

Formula: C₄H₆KNO₄·xH₂O
MW: 171.19 g/mol (anhy)
MDL Number: MFCD00021083
CAS Number: 1115-63-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c2380d44392d7831877bbe99263816e39b33d93&vid=334980926393dc2fe080d4f7eb1ff2dff34ba793&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR