Diphenylsulfone-3,3'-disulfonic acid disodium salt ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Disodium Diphenylsulphone-3,3'-disulphonate, Disodium Diphenylsulfone-3,3'-disulfonate

D0916-5G D0916-25G
TCD0916-5GEA 282.82 CAD
TCD0916-5G TCD0916-025G
Diphenylsulfone-3,3'-disulfonic acid disodium salt ≥97.0% (by titrimetric analysis)
Diphenylsulfone-3,3'-disulfonic acid disodium salt

CAS Number: 39616-93-8
MDL Number: MFCD00059845
Molecular Formula: C12H10O8S3
Molecular Weight: 422.35
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Diphenyl Sulfones (for High-Performance Polymer Research)
Other Product Names / Synonyms: Disodium Diphenylsulfone-3,3'-disulfonate

Formula: C₁₂H₁₀O₈S₃
MW: 422.35 g/mol
MDL Number: MFCD00059845
CAS Number: 39616-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11455ca09ac6928c0942677780ee257a705af234&vid=11455ca09ac6928c0942677780ee257a705af234&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR