trans,trans-Dibenzylideneacetone ≥99.0%

Supplier: TCI America

D0903-250G D0903-25G
TCD0903-250GEA 595.66 CAD
TCD0903-250G TCD0903-025G
trans,trans-Dibenzylideneacetone ≥99.0%
trans,trans-Dibenzylideneacetone

CAS Number: 35225-79-7
MDL Number: MFCD00004790
Molecular Formula: C17H14O
Molecular Weight: 234.30
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 113


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: trans,trans-Dibenzalacetone, trans,trans-Dibenzylideneacetone, trans,trans-Distyryl Ketone

Formula: C₁₇H₁₄O
Melting Pt: 113 °C
MDL Number: MFCD00004790
CAS Number: 35225-79-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd2914f75cc79bdf2a73ad94063d865509ebdbbd&vid=dd2914f75cc79bdf2a73ad94063d865509ebdbbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR