2,4-Diphenyloxazole ≥99.0%

Supplier: TCI America

D0901-25G D0901-1G
TCD0901-025GEA 552.05 CAD
TCD0901-025G TCD0901-001G
2,4-Diphenyloxazole ≥99.0%
2,4-Diphenyloxazole

CAS Number: 838-41-5
MDL Number: MFCD00016890
Molecular Formula: C15H11NO
Molecular Weight: 221.26
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 340
Melting point (°C): 105

Formula: C₁₅H₁₁NO
Boiling Pt: 340 °C
Melting Pt: 105 °C
MDL Number: MFCD00016890
CAS Number: 838-41-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18174af913c9b1471b964423ea977323b2f574b9&vid=18174af913c9b1471b964423ea977323b2f574b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR