Di(alpha-phenylethyl) sulfide (DL and meso mixture)

Supplier: TCI America

D0889-5G D0889-25G
TCD0889-005GEA 272.08 CAD
TCD0889-005G TCD0889-025G
Di(alpha-phenylethyl) sulfide (DL and meso mixture)
Di(alpha-phenylethyl) sulfide (DL and meso mixture)

CAS Number: 838-59-5
MDL Number: MFCD00026342
Molecular Formula: C16H18S
Molecular Weight: 242.38
Form: Clear Liquid
Color: Very Pale Yellow


Other Product Names / Synonyms: 1-Phenylethyl Sulfide

Formula: C₁₆H₁₈S
MDL Number: MFCD00026342
CAS Number: 838-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b66a746a7470cf6fc482cfa191fb59454fb5041&vid=9b66a746a7470cf6fc482cfa191fb59454fb5041&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR