Diphenylamine sulfate ≥98.0%

Supplier: TCI America

D0875-500G D0875-25G
TCD0875-500GEA 881.64 CAD
TCD0875-500G TCD0875-025G
Diphenylamine sulfate ≥98.0%
Diphenylamine sulfate

CAS Number: 587-84-8
MDL Number: MFCD00042016
Molecular Formula: C12H11N
Molecular Weight: 267.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150

Formula: C₁₂H₁₁N
Melting Pt: 150 °C
MDL Number: MFCD00042016
CAS Number: 587-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d44130126ad9e22f3c044c6a71ace8dc07252ffa&vid=d44130126ad9e22f3c044c6a71ace8dc07252ffa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR