Disodium-4,4'-dinitrostilbene-2,2'-disulfonate ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Dinitrostilbene-2,2'-disulfonic Acid Disodium Salt, 4,4'-Dinitrostilbene-2,2'-disulphonic Acid Disodium Salt
D0853-25G
D0853-500G
D0853-100G
TCD0853-025GEA
52.14
CAD
TCD0853-025G
TCD0853-500G
TCD0853-100G
Disodium-4,4'-dinitrostilbene-2,2'-disulfonate ≥98.0%
Disodium-4,4'-dinitrostilbene-2,2'-disulfonate
CAS Number: 3709-43-1
MDL Number: MFCD00024929
Molecular Formula: C14H10N2O10S2
Molecular Weight: 474.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 4,4'-Dinitrostilbene-2,2'-disulfonic Acid Disodium Salt
Formula:
C₁₄H₁₀N₂O₁₀S₂ MW: 474.32 g/mol |
MDL Number:
MFCD00024929 CAS Number: 3709-43-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...