Bis(4-nitrophenyl)methane ≥99.0%

Supplier: TCI America

D0833-1G D0833-5G
TCD0833-001GEA 148.84 CAD
TCD0833-001G TCD0833-005G
Bis(4-nitrophenyl)methane ≥99.0%
Bis(4-nitrophenyl)methane

CAS Number: 1817-74-9
MDL Number: MFCD00059184
Molecular Formula: C13H10N2O4
Molecular Weight: 258.23
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 190

Formula: C₁₃H₁₀N₂O₄
Melting Pt: 190 °C
MDL Number: MFCD00059184
CAS Number: 1817-74-9
EINECS: 217-330-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3128661f096f3445928f8cb29049ce8342a5e5aa&vid=3128661f096f3445928f8cb29049ce8342a5e5aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR