4,4'-Dinitrodiphenyl ether ≥99.0%
Supplier: TCI America
Synonyms:
4,4'-Dinitrodiphenyl Oxide
D0832-10G
D0832-1G
TCD0832-010GEA
85.32
CAD
TCD0832-010G
TCD0832-001G
4,4'-Dinitrodiphenyl ether ≥99.0%
4,4'-Dinitrodiphenyl ether
CAS Number: 101-63-3
MDL Number: MFCD00059182
Molecular Formula: C12H8N2O5
Molecular Weight: 260.21
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 144
Product Categories: Materials Chemistry, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Dinitrodiphenyl Oxide
Formula:
C₁₂H₈N₂O₅ MW: 260.21 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD00059182 CAS Number: 101-63-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...