N,N'-Di-2-naphthyl-1,4-phenylenediamine ≥96.0%

Supplier: TCI America

Synonyms: N,N-Di-2-naphthyl-1,4-phenylenediamine

D0812-25G D0812-500G
TCD0812-025GEA 50.56 CAD
TCD0812-025G TCD0812-500G
N,N'-Di-2-naphthyl-1,4-phenylenediamine ≥96.0%
N,N'-Di-2-naphthyl-1,4-phenylenediamine

CAS Number: 93-46-9
MDL Number: MFCD00053487
Molecular Formula: C26H20N2
Molecular Weight: 360.46
Purity/Analysis Method: >96.0% (N)
Form: Crystal
Flash Point (°C): 300

Formula: C₂₆H₂₀N₂
MW: 360.46 g/mol
Flash Pt: 300 °C
MDL Number: MFCD00053487
CAS Number: 93-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e9ef63b092c03250e38862b05dc01dcbd2e61f3&vid=8e9ef63b092c03250e38862b05dc01dcbd2e61f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR