1,3-Di(p-tolyl)-2-thiourea ≥98.0% (by HPLC)
Supplier: TCI America
D0803-5G
D0803-25G
TCD0803-005GEA
120.08
CAD
TCD0803-005G
TCD0803-025G
1,3-Di(p-tolyl)-2-thiourea ≥98.0% (by HPLC)
1,3-Di(p-tolyl)-2-thiourea
CAS Number: 621-01-2
MDL Number: MFCD00008541
Molecular Formula: C15H16N2S
Molecular Weight: 256.37
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 179
Formula:
C₁₅H₁₆N₂S Melting Pt: 179 °C |
MDL Number:
MFCD00008541 CAS Number: 621-01-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...