3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D0801-1G D0801-5G
TCD0801-001GEA 177.75 CAD
TCD0801-001G TCD0801-005G
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98.0% (by titrimetric analysis)
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

CAS Number: 298-93-1
MDL Number: MFCD00011964
Molecular Formula: C18H16BrN5S
Molecular Weight: 414.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 195


Product Categories: Bioscience, Stains & Dyes, Redox Research Reagents
Other Product Names / Synonyms: MTT, Thiazolyl Blue Tetrazolium Bromide

Formula: C₁₈H₁₆N₅BrS
Melting Pt: 195 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=232747b2c66a62464fdd80013c8e205b94efd442&vid=232747b2c66a62464fdd80013c8e205b94efd442&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR