N,N-Dimethyl-2-naphthylamine ≥98.0%

Supplier: TCI America

Synonyms: N,N-Dimethylnaphthalen-2-amine, 2-Dimethylaminonaphthalene

D0756-1G D0756-5G
TCD0756-001GEA 26.86 CAD
TCD0756-001G TCD0756-5G
N,N-Dimethyl-2-naphthylamine ≥98.0%
N,N-Dimethyl-2-naphthylamine

CAS Number: 2436-85-3
MDL Number: MFCD00059550
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 305
Melting point (°C): 46


Other Product Names / Synonyms: 2-Dimethylaminonaphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 305 °C
Melting Pt: 46 °C
MDL Number: MFCD00059550
CAS Number: 2436-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22126cf3c90620e264aa13e23dacd85c1b9c154b&vid=22126cf3c90620e264aa13e23dacd85c1b9c154b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR