3,5-Dimethylcyclohexanol (mixture of isomers) ≥98.0%
Supplier: TCI America
D0702-25ML
D0702-500ML
TCD0702-025MLEA
148.2
CAD
TCD0702-025ML
TCD0702-500ML
3,5-Dimethylcyclohexanol (mixture of isomers) ≥98.0%
3,5-Dimethylcyclohexanol (mixture of isomers)
CAS Number: 5441-52-1
MDL Number: MFCD00001447
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 186
Flash Point (°C): 73
Freezing point (°C): 29
Formula:
C₈H₁₆O Boiling Pt: 186 °C Flash Pt: 73 °C |
MDL Number:
MFCD00001447 CAS Number: 5441-52-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...