2,5-Dimethyl-p-benzoquinone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,5-Dimethylcyclohexa-2,5-diene-1,4-dione
D0686-5G
D0686-25G
D0686-1G
TCD0686-005GEA
181.7
CAD
TCD0686-005G
TCD0686-025G
TCD0686-1G
2,5-Dimethyl-p-benzoquinone ≥98.0% (by GC, titration analysis)
2,5-Dimethyl-p-benzoquinone
CAS Number: 137-18-8
MDL Number: MFCD00041737
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125
Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction
Other Product Names / Synonyms: 2,5-Dimethyl-1,4-benzoquinone, 2,5-Dimethyl-p-quinone, Phlorone
Formula:
C₈H₈O₂ MW: 136.15 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00041737 CAS Number: 137-18-8 EINECS: 205-283-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...