Sodium-2,4-dimethylbenzenesulfonate monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0682-25G D0682-500G
TCD0682-025GEA 58.46 CAD
TCD0682-025G TCD0682-500G
Sodium-2,4-dimethylbenzenesulfonate monohydrate ≥98.0% (by titrimetric analysis)
Sodium-2,4-dimethylbenzenesulfonate monohydrate

CAS Number: 142063-30-7
MDL Number: MFCD00041883
Molecular Formula: C8H10O3S
Molecular Weight: 208.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2,4-Dimethylbenzenesulfonic Acid Sodium Salt, Sodium m-Xylene-4-sulfonate, m-Xylene-4-sulfonic Acid Sodium Salt

Formula: C₈H₁₀O₃S
MDL Number: MFCD00041883
CAS Number: 142063-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0c594a3b45055137fa32b377a8af31be254a7cc&vid=438cc84fe9b7e483da78b855ac7ca33bfb432efc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR