3,3'-Dimethylazobenzene ≥98.0%

Supplier: TCI America

D0676-1G D0676-5G
TCD0676-001GEA 129.56 CAD
TCD0676-001G TCD0676-005G
3,3'-Dimethylazobenzene ≥98.0%
3,3'-Dimethylazobenzene

CAS Number: 588-04-5
MDL Number: MFCD00048080
Molecular Formula: C14H14N2
Molecular Weight: 210.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 53


Other Product Names / Synonyms: m-Azotoluene

Formula: C₁₄H₁₄N₂
Melting Pt: 53 °C
MDL Number: MFCD00048080
CAS Number: 588-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a900c2d0928de595b845e97b3d8d4daf53243407&vid=a900c2d0928de595b845e97b3d8d4daf53243407&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR