3-Chloropropyldimethylammonium chloride ≥98.0% for synthesis

Supplier: TCI America

Synonyms: 3-Dimethylaminopropylchloride hydrochloride, 3-Dimethylaminopropyl chloride hydrochloride, N-(3-Chloropropyl)-N,N-dimethylammonium chloride, 3-Chloro-N,N-dimethylpropan-1-amine hydrochloride

D0663-25G D0663-250G
TCD0663-025GEA 31.6 CAD
TCD0663-025G TCD0663-250G
3-Chloropropyldimethylammonium chloride ≥98.0% for synthesis
3-Chloropropyldimethylammonium chloride

CAS Number: 5407-04-5
MDL Number: MFCD00012521
Molecular Formula: C5H12ClN
Molecular Weight: 158.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 143


Other Product Names / Synonyms: 3-Chloro-N,N-dimethylpropylamine Hydrochloride

Formula: C₅H₁₃Cl₂N
MW: 158.07 g/mol
Melting Pt: 143 °C
MDL Number: MFCD00012521
CAS Number: 5407-04-5
EINECS: 226-467-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5b1353d9172a49a6f293cf269eff80745d475f6&vid=d5b1353d9172a49a6f293cf269eff80745d475f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR