Sodium-6,7-dihydroxynaphthalene-2-sulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dihydroxynaphthalene-6-sulphonic acid Monosodium Salt, DS acid Monosodium Salt, Monosodium 6,7-Dihydroxynaphthalene-2-sulfonate, Sodium 2,3-dihydroxynaphthalene-6-sulphonate, Monosodium 6,7-Dihydroxynaphthalene-2-sulphonate, Sodium 2,3-dihydroxynaphthalene-6-sulfonate, 2,3-Dihydroxynaphthalene-6-sulfonic acid Monosodium Salt

D0597-25G D0597-500G
TCD0597-025GEA 127.98 CAD
TCD0597-025G TCD0597-500G
Sodium-6,7-dihydroxynaphthalene-2-sulfonate ≥98.0% (by HPLC, titration analysis)
Sodium-6,7-dihydroxynaphthalene-2-sulfonate

CAS Number: 135-53-5
MDL Number: MFCD00004074
Molecular Formula: C10H8O5S
Molecular Weight: 262.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: DS Acid Monosodium Salt, Monosodium 6,7-Dihydroxynaphthalene-2-sulfonate, 2,3-Dihydroxynaphthalene-6-sulfonic Acid Monosodium Salt

Formula: C₁₀H₈O₅S
MW: 262.21 g/mol
MDL Number: MFCD00004074
CAS Number: 135-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c421174704cbecd24caaa1f9b642132587b3373&vid=4c421174704cbecd24caaa1f9b642132587b3373&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR