3,6-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Disodium 2,7-Dihydroxynaphthalene-3,6-disulphonate, 3,6-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt, 3,6-Dihydroxy-2,7-naphthalenedisulfonic acid disodium salt, Disodium 3,6-dihydroxynaphthalene-2,7-disulfonate, Sodium 3,6-dihydroxynaphthalene-2,7-disulphonate, Disodium 3,6-dihydroxynaphthalene-2,7-disulphonate, 2,7-Dihydroxynaphthalene-3,6-disulfonic acid disodium salt, Sodium 3,6-dihydroxynaphthalene-2,7-disulfonate, Disodium 2,7-Dihydroxynaphthalene-3,6-disulfonate, 2,7-Dihydroxynaphthalene-3,6-disulphonic acid disodium salt

D0595-500G D0595-25G
TCD0595-500GEA 584.6 CAD
TCD0595-500G TCD0595-025G
3,6-Dihydroxynaphthalene-2,7-disulfonic acid disodium salt ≥98.0% for synthesis
3,6-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt

CAS Number: 7153-21-1
MDL Number: MFCD00004090
Molecular Formula: C10H8O8S2
Molecular Weight: 364.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2,7-Dihydroxynaphthalene-3,6-disulfonic Acid Disodium Salt, Disodium 3,6-Dihydroxynaphthalene-2,7-disulfonate, Disodium 2,7-Dihydroxynaphthalene-3,6-disulfonate

Formula: C₁₀H₆Na₂O₈S₂
MDL Number: MFCD00004090
CAS Number: 7153-21-1
EINECS: 230-499-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3421f83d74688d52905fe7b185e314c171e6651&vid=d3421f83d74688d52905fe7b185e314c171e6651&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR