1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate ≥98.0% for synthesis

Supplier: TCI America
Warning

Synonyms: Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulfonate monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate, Catechol-3,5-disulfonic acid disodium salt monohydrate, 3,5-Pyrocatecholdisulfonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulfonic acid disodium salt monohydrate

D0567-250G D0567-25G
TCD0567-250GEA 358.66 CAD
TCD0567-250G TCD0567-025G
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate ≥98.0% for synthesis
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate

CAS Number: 149-45-1
MDL Number: MFCD00007473
Molecular Formula: C6H6O8S2
Molecular Weight: 314.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Catechol-3,5-disulfonic Acid Disodium Salt, Disodium 4,5-Dihydroxybenzene-1,3-disulfonate, Pyrocatechol-3,5-disulfonic Acid Disodium Salt, Tiferron

Formula: C₆H₄Na₂O₈S₂·1H₂O
MW: 332.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149531
CAS Number: 270573-71-2
EINECS: 205-741-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee00b8039b1e2258dfe079e5730461ccfbf3a18e&vid=ee00b8039b1e2258dfe079e5730461ccfbf3a18e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR