3,5-Diethoxycarbonyl-1,4-dihydro-2,4,6-collidine ≥98.0%

Supplier: TCI America

Synonyms: DDC, 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Diethyl 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate

D0558-1G D0558-5G D0558-25G
TCD0558-001GEA 26.86 CAD
TCD0558-001G TCD0558-5G TCD0558-25G
3,5-Diethoxycarbonyl-1,4-dihydro-2,4,6-collidine ≥98.0%
3,5-Diethoxycarbonyl-1,4-dihydro-2,4,6-collidine

CAS Number: 632-93-9
MDL Number: MFCD00005950
Molecular Formula: C14H21NO4
Molecular Weight: 267.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 132


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models
Other Product Names / Synonyms: 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 1,4-Dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, DDC

Formula: C₁₄H₂₁NO₄
MW: 267.33 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00005950
CAS Number: 632-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9832684697237ebcb5ae86e878f526cb994c1567&vid=9832684697237ebcb5ae86e878f526cb994c1567&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR