Butopyronoxyl ≥95.0% (by HPLC)

Supplier: TCI America

D0550-25G D0550-500G
TCD0550-025GEA 124.82 CAD
TCD0550-025G TCD0550-500G
Butopyronoxyl ≥95.0% (by HPLC)
Butopyronoxyl

CAS Number: 532-34-3
MDL Number: MFCD00006574
Molecular Formula: C12H18O4
Molecular Weight: 226.27
Purity/Analysis Method: >95.0% (HPLC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 157
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Butyl 3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylate, 3,4-Dihydro-2,2-dimethyl-4-oxo-2H-pyran-6-carboxylic Acid Butyl Ester, Indalone

Formula: C₁₂H₁₈O₄
Boiling Pt: 113 °C
Density: 1.06 (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00006574
CAS Number: 532-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3f5d558f970b1d42cb62bf9795fcd05af18f716&vid=c3f5d558f970b1d42cb62bf9795fcd05af18f716&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR