N,N-Diethyl-o-toluidine ≥98.0%
Supplier: TCI America
Synonyms:
N,N-Diethyl-2-methylaniline, 2-Diethylaminotoluene
D0531-5ML
D0531-25ML
TCD0531-5MLEA
158
CAD
TCD0531-5ML
TCD0531-025ML
N,N-Diethyl-o-toluidine ≥98.0%
N,N-Diethyl-o-toluidine
CAS Number: 606-46-2
MDL Number: MFCD00059227
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Melting point (°C): -60
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: 2-Diethylaminotoluene, N,N-Diethyl-2-methylaniline
Formula:
C₁₁H₁₇N MW: 163.26 g/mol Boiling Pt: 210 °C Melting Pt: –60 °C Density: 0.91 (20 °C) |
MDL Number:
MFCD00059227 CAS Number: 606-46-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...