N,N-Diethyl-1,4-phenylenediamine dihydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Amino-N,N-diethylaniline Dihydrochloride

D0519-25G D0519-5G
TCD0519-025GEA 249.64 CAD
TCD0519-025G TCD0519-5G
N,N-Diethyl-1,4-phenylenediamine dihydrochloride ≥98.0% (by titrimetric analysis)
N,N-Diethyl-1,4-phenylenediamine dihydrochloride

CAS Number: 16713-15-8
MDL Number: MFCD00060227
Molecular Formula: C10H16N2
Molecular Weight: 237.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4-Amino-N,N-diethylaniline Dihydrochloride

Formula: C₁₀H₁₆N₂
MW: 237.17 g/mol
MDL Number: MFCD00060227
CAS Number: 16713-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=881568c464ffcdebea23a18b876a84ffaa8be03d&vid=881568c464ffcdebea23a18b876a84ffaa8be03d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR