N,N-Diethyl-N'-oleoylethylenediamine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D0517-5G D0517-25G
TCD0517-005GEA 55.3 CAD
TCD0517-005G TCD0517-25G
N,N-Diethyl-N'-oleoylethylenediamine ≥97.0% (by titrimetric analysis)
N,N-Diethyl-N'-oleoylethylenediamine

CAS Number: 13282-67-2
MDL Number: MFCD00027344
Molecular Formula: C24H48N2O
Molecular Weight: 380.66
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: N'-Oleoyl-N,N-diethylethylenediamine

Formula: C₂₄H₄₈N₂O
Density: 0.89 (20 °C)
MDL Number: MFCD00027344
CAS Number: 13282-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d46006c50ce1d8653bb664fce127ccee811ec1&vid=b0d46006c50ce1d8653bb664fce127ccee811ec1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR