Cinnamyl acetate ≥97.0%

Supplier: TCI America

Synonyms: Acetic Acid Cinnamyl Ester

A0901-25ML A0901-500ML
TCA0901-025MLEA 45.82 CAD
TCA0901-025ML TCA0901-500ML
Cinnamyl acetate ≥97.0%
Cinnamyl acetate

CAS Number: 103-54-8
MDL Number: MFCD00008722
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 265
Flash Point (°C): 118
Specific Gravity (20/20): 1.05


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: Acetic Acid Cinnamyl Ester

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 265 °C
Density: 1.05 (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD00008722
CAS Number: 103-54-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a38bf282eb98dec22d2b45ffc9b91a26a5601a1d&vid=a38bf282eb98dec22d2b45ffc9b91a26a5601a1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR