N,N-Diethylglycinesodium salt ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D0509-25G D0509-100G
TCD0509-025GEA 230.68 CAD
TCD0509-025G TCD0509-100G
N,N-Diethylglycinesodium salt ≥97.0% (by titrimetric analysis)
N,N-Diethylglycinesodium salt

CAS Number: 5426-55-1
MDL Number: MFCD00053497
Molecular Formula: C6H13NO2
Molecular Weight: 153.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Formula: C₆H₁₃NO₂
MDL Number: MFCD00053497
CAS Number: 5426-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8b541ddf5dbe504004f6e6ee33c0fb5be8637a2&vid=f8b541ddf5dbe504004f6e6ee33c0fb5be8637a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR