Hexestrol ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-(1,2-Diethylethylene)diphenol, rel-4,4'-((3R,4S)-Hexane-3,4-diyl)diphenol

D0494-10G D0494-1G
TCD0494-010GEA 413.96 CAD
TCD0494-010G TCD0494-001G
Hexestrol ≥98.0%
Hexestrol

CAS Number: 84-16-2
MDL Number: MFCD00068996
Molecular Formula: C18H22O2
Molecular Weight: 270.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 186


Other Product Names / Synonyms: 4,4'-(1,2-Diethylethylene)diphenol

Formula: C₁₈H₂₂O₂
MW: 270.37 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00068996
CAS Number: 84-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66c43f981c55c307314193f24481927375151244&vid=66c43f981c55c307314193f24481927375151244&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR