Diethylammonium diethyldithiocarbamate ≥97.0%

Supplier: TCI America

Synonyms: Diethyldithiocarbamic Acid Diethylammonium Salt, Diethylammonium diethylcarbamodithioate

D0488-25G D0488-500G
TCD0488-025GEA 186.44 CAD
TCD0488-025G TCD0488-500G
Diethylammonium diethyldithiocarbamate ≥97.0%
Diethylammonium diethyldithiocarbamate

CAS Number: 1518-58-7
MDL Number: MFCD00013146
Molecular Formula: C5H11NS2
Molecular Weight: 222.41
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 83


Other Product Names / Synonyms: Diethyldithiocarbamic Acid Diethylammonium Salt

Formula: C₅H₁₁NS₂
MW: 222.41 g/mol
Melting Pt: 83 °C
MDL Number: MFCD00013146
CAS Number: 1518-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99b1500a922016c30988b919527debeac53f37ae&vid=99b1500a922016c30988b919527debeac53f37ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR