Copper(II) diethyldithiocarbamate ≥97.0%

Supplier: TCI America

Synonyms: Diethyldithiocarbamic Acid Copper(II) Salt

D0487-25G D0487-5G
TCD0487-025GEA 284.4 CAD
TCD0487-025G TCD0487-5G
Copper(II) diethyldithiocarbamate ≥97.0%
Copper(II) diethyldithiocarbamate

CAS Number: 13681-87-3
MDL Number: MFCD00070506
Molecular Formula: C5H11NS2
Molecular Weight: 360.07
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 197


Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Diethyldithiocarbamic Acid Copper(II) Salt

Formula: C₅H₁₁NS₂
MW: 360.07 g/mol
Melting Pt: 197 °C
MDL Number: MFCD00070506
CAS Number: 13681-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20f7949b13cff0070d28ac0a7acd7496b169262f&vid=20f7949b13cff0070d28ac0a7acd7496b169262f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR