Ethyl arachidate ≥95.0%

Supplier: TCI America

A0899-25G A0899-5G
TCA0899-025GEA 909.27 CAD
TCA0899-025G TCA0899-005G
Ethyl arachidate ≥95.0%
Ethyl arachidate

CAS Number: 18281-05-5
MDL Number: MFCD00056233
Molecular Formula: C22H44O2
Molecular Weight: 340.59
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 44


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Icosanoic Acid Ethyl Ester, Arachidic Acid Ethyl Ester, Ethyl Icosanoate

Formula: C₂₂H₄₄O₂
Boiling Pt: 215 °C
Melting Pt: 44 °C
MDL Number: MFCD00056233
CAS Number: 18281-05-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05b3c0a2ebe80ecd93d64aaa82332c9eda33f189&vid=05b3c0a2ebe80ecd93d64aaa82332c9eda33f189&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR