N-(2-Diethylaminoethyl)-1-naphthylamine oxalate 98,0
Supplier: TCI America
Synonyms:
N,N-Diethyl-N'-1-naphthylethylenediamine Oxalate, N1,N1-Diethyl-N2-(naphthalen-1-yl)ethane-1,2-diamine oxalate, Tsuda's Reagent
D0469-5G
D0469-1G
D0469-25G
TCD0469-005GEA
125.4
CAD
TCD0469-005G
TCD0469-001G
TCD0469-25G
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate 98,0
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate
CAS Number: 29473-53-8
MDL Number: MFCD00060156
Molecular Formula: C16H22N2
Molecular Weight: 332.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: N,N-Diethyl-N'-1-naphthylethylenediamine Oxalate, Tsuda's Reagent
Formula:
C₁₆H₂₄N₂O₄ MW: 332.40 g/mol |
MDL Number:
MFCD00060156 CAS Number: 29473-53-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...